missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
140.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Color | Yellow |
Packaging | Glass bottle |
Assay Percent Range | 47.5% min. (Mo) |
Quantity | 500 g |
Linear Formula | H3[P(Mo3O10)4]·xH2O |
Physical Form | Crystals |
Merck Index | 14, 7342 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and ether |
Formula Weight | 1825.25 |
Percent Purity | 80% |
Show More |
Safety and Handling
Product Identifier
- Phosphomolybdic acid hydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'