Learn More
Alfa Aesar™ Phosphoenolpyruvic acid tricyclohexylammonium salt
Metabolic intermediate in many kinase reactions
Brand: Alfa Aesar™ J61943.06
Code : NEW
Additional Details : CAS Number : 35556-70-8 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
Phosphoenolpyruvic acid tricyclohexylammonium salt | |
C3H5O6P·3C6H13N | |
MFCD00004258 | |
2-(Phosphonooxy)-2-propenoic acid tri(cyclohexylammonium) salt; PEP-tri |
35556-70-8 | |
465.58 | |
3752336 |
Specifications
Phosphoenolpyruvic acid tricyclohexylammonium salt | |
Powder | |
C3H5O6P·3C6H13N | |
MFCD00004258 | |
2-(Phosphonooxy)-2-propenoic acid tri(cyclohexylammonium) salt; PEP-tri | |
5g |
35556-70-8 | |
≥99% | |
465.58 | |
3752336 | |
White |