missing translation for 'onlineSavingsMsg'
Learn More

Phospho-ZAP70/Syk (Tyr319, Tyr352), PerCP-eFluor™ 710, clone: n3kobu5, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-9006-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15539386

  • £224.78 / 100 tests


Estimated Shipment date
from Supplier 28-06-2018
Add to basket

Description

Description

Description: This n3kobu5 monoclonal antibody recognizes human and mouse zeta chain-associated protein of 70 kD (also known as ZAP-70) and spleen tyrosine kinase (also known as SYK) when phosphorylated on Y319 and Y352, respectively. ZAP-70 and SYK are members of the SYK protein tyrosine kinase (PTK) family that are phosphorylated and activated by Src family PTKs. ZAP-70/SYK Y319/Y352 are located in the so-called interdomain of ZAP-70/SYK (between the N-terminal dual SH2 domains and the C-terminal kinase domain).Phosphorylation of ZAP-70 Y319 by Lck is necessary for T cell receptor (TCR)-dependent association of ZAP-70 with Lck and phospholipase C gamma and subsequent activation of calcium-dependent and Ras signaling cascades. SYK Y352 phosphorylation by Fyn/Lyn is critical for propagation of B cell receptor (BCR) signaling and for B cell development.Specificity of this n3kobu5 clone was determined by ELISA, flow cytometry, and western blotting.Applications Reported: This n3kobu5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This n3kobu5 antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Staining Protocol: All protocols work well for this monoclonal antibody. Use of Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins allows for the greatest flexibility for detection of surface and intracellular (cytoplasmic) proteins. Use of Protocol B: One-step protocol: intracellular (nuclear) proteins is recommended for staining of transcription factors in conjunction with surface and phosphorylated intracellular (cytoplasmic) proteins. Protocol C: Two-step protocol: Fixation/Methanol allows for the greatest discrimination of phospho-specific signaling between unstimulated and stimulated samples, but with limitations on the ability to stain specific surface proteins (refer to Clone Performance Following Fixation/Permeabilization located in the Best Protocols Section under the Resources tab online). All Protocols can be found in the Flow Cytometry Protocols: Staining Intracellular Antigens for Flow Cytometry Protocol located in the Best Protocols Section under the Resources tab online.PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Phospho-ZAP-70/SYK (Y319/Y352)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P43404, P48025, P43405, P43403
Zeta chain-associated protein of 70 kD/Spleen tyrosine kinase
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
HumanĂ¾Murine
Flow Cytometry
n3kobu5
PerCP-eFluor 710
SYK
Conjugated
SYK, Zap70
IgG2b, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
6850, 7535
Videos

For Research Use Only.