Learn More
phospho-Tyrosine Monoclonal Antibody (pY20), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£111.00 - £295.00
Specifications
Clone | pY20 |
---|---|
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PerCP-eFluor 710 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15569256
|
Affymetrix eBioscience
46-5001-42 |
100 Tests |
£295.00
100 tests |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15559256
|
Affymetrix eBioscience
46-5001-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The role of tyrosine phosphorylation in transduction of the mitogenic signal from transmembrane receptors and in transformation by oncogene tyrosine kinases has been the subject of intense investigation for several years. While the phosphorylation of specific tyrosine residues has been shown to be a primary mechanism of signal transduction during normal mitogenesis, cell cycle progression and oncogenic transformation, its role in other areas such as differentiation and gap junction communication, is a matter of active and ongoing research. Antibodies that specifically recognize phosphorylated tyrosine residues have proved to be invaluable to the study of tyrosine -phosphorylated proteins and the biochemical pathways in which they function. The fluorescein (FITC) conjugate of clone PY20 anti-phosphotyrosine is especially useful for the detection of these P-Tyr proteins in immunohistochemical and immunocytochemical protocols in situations wherein the use of a secondary antibody would complicate detection of the protein(s) of interest.
phospho-Tyrosine Monoclonal antibody specifically detects phospho-Tyrosine in Chemical samples. It is validated for Flow Cytometry.Specifications
pY20 | |
Monoclonal | |
Liquid | |
RUO | |
Primary | |
Antibody |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.