Learn More
Phospho-TRIM28 (Ser824) Recombinant Polyclonal Antibody, Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 711116
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with Monkey, Mouse, Rat Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Phospho-TRIM28 (Ser824) Recombinant Polyclonal antibody specifically detects Phospho-TRIM28 (Ser824) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
Phospho-TRIM28 (Ser824) | |
Recombinant Polyclonal | |
Unconjugated | |
TRIM28 | |
AA408787; E3 SUMO-protein ligase TRIM28; KAP1; KAP-1; KRAB [Kruppel-associated box domain]-associated protein 1; KRAB-A-interacting protein; KRAB-associated protein 1; KRAB-interacting p; KRAB-interacting protein 1; Krip1; KRIP-1; MommeD9; Nuclear corepressor KAP-1; PPP1R157; protein phosphatase 1, regulatory subunit 157; RING finger protein 96; RING-type E3 ubiquitin transferase TIF1-beta; RNF96; TF1B; tif1 beta; Tif1b; Tif1beta; TIF1-beta; Transcription intermediary factor 1-beta; transcriptional intermediary factor 1, beta; transcriptional intermediary factor 1-beta; Trim28; tripartite motif; tripartite motif containing 28; tripartite motif protein 28; tripartite motif-containing 28; tripartite motif-containing protein 28 | |
Rabbit | |
Protein A | |
RUO | |
10155 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q13263 | |
TRIM28 | |
Peptide corresponding to human TRIM28 (Ser824) [aa819-aa827] | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |