missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-TOP2A (Ser1213) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564536
Additional Details : Weight : 0.01000kg
Description
Phospho-TOP2A (Ser1213) Polyclonal Antibody detects endogenous levels of TOP2A only when phosphorylated at Ser1213.
Phospho-TOP2A (Ser1213) Polyclonal antibody specifically detects Phospho-TOP2A (Ser1213) in Human, Mouse samples. It is validated for Western Blot.Specifications
Phospho-TOP2A (Ser1213) | |
Polyclonal | |
Unconjugated | |
TOP2A | |
DNA gyrase; DNA topoisomerase (ATP-hydrolyzing); DNA topoisomerase 2-alpha; DNA topoisomerase 2-alpha-like protein; DNA Topoisomerase II alpha; DNA topoisomerase II, 170 kD; DNA topoisomerase II, alpha isozyme; EC 5.99.1.3; OTTHUMP00000207108; P11388-1; TOP2; Top-2; Top2a; TOPOII; topoisomerase (DNA) 2 alpha; topoisomerase (DNA) II alpha; topoisomerase (DNA) II alpha 170kDa; topoisomerase ii alpha; topoisomersae II; TP2A | |
Rabbit | |
Affinity chromatography | |
RUO | |
21973, 7153 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P11388, Q01320 | |
TOP2A | |
A synthesized peptide derived from human TOP2A (Phospho-Ser1213) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |