missing translation for 'onlineSavingsMsg'
Learn More

Phospho-STAT6 (Tyr641), PerCP-eFluor™ 710, clone: CHI2S4N, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-9013-42

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15599386

  • £228.00 / 100 tests

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This CHI2S4N monoclonal antibody recognizes human and mouse signal transducer and activator of transcription 6 (STAT6) when phosphorylated on tyrosine 641. Following their phosphorylation by JAKs, STAT proteins translocate to the nucleus where they bind to DNA and regulate transcription of specific genes in a cell type- and cytokine-specific manner. In response to IL-4, STAT6 is phosphorylated on tyrosine 641 by JAK1 and JAK3. STAT6 signaling downstream of the IL-4 receptor promotes T cell growth and B cell production of IgE. Specificity of this CHI2S4N clone was determined by ELISA and flow cytometry.Applications Reported: This CHI2S4N antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This CHI2S4N antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Staining Protocol: We recommend using Protocol C: Two-step protocol: Fixation/Methanol. Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins and Protocol B: One-step protocol: intracellular (nuclear) proteins cannot be used. All Protocols can be found in the Flow Cytometry Protocols: Staining Intracellular Antigens for Flow Cytometry Protocol located in the Best Protocols Section under the Resources tab online. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Phospho-STAT6 (Tyr641)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P52633, P42226
Signal Transducers and Activators of Transcription 6, IL4 STAT, Interleukin 4 Induced
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Murine
Flow Cytometry
CHI2S4N
PerCP-eFluor 710
STAT6
Liquid
Stat6
IgG2a, kappa, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
20852, 6778
Videos
SDS
Documents

For Research Use Only.