missing translation for 'onlineSavingsMsg'
Learn More

Phospho-STAT6 (Tyr641), PE, clone: CHI2S4N, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9013-41

Code : Z2

Additional Details : Weight : 0.00250kg

 View more versions of this product

Product Code. 15202635

  • £80.50 / 25 tests


EU Stock 3
Add to basket

Description

Description

Description: This CHI2S4N monoclonal antibody recognizes human and mouse signal transducer and activator of transcription 6 (STAT6) when phosphorylated on tyrosine 641. Following their phosphorylation by JAKs, STAT proteins translocate to the nucleus where they bind to DNA and regulate transcription of specific genes in a cell type- and cytokine-specific manner. In response to IL-4, STAT6 is phosphorylated on tyrosine 641 by JAK1 and JAK3. STAT6 signaling downstream of the IL-4 receptor promotes T cell growth and B cell production of IgE. Specificity of this CHI2S4N clone was determined by ELISA and flow cytometry.Applications Reported: This CHI2S4N antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This CHI2S4N antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Staining Protocol: We recommend using Protocol C: Two-step protocol: Fixation/Methanol. Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins and Protocol B: One-step protocol: intracellular (nuclear) proteins cannot be used. All Protocols can be found in the Flow Cytometry Protocols: Staining Intracellular Antigens for Flow Cytometry Protocol located in the Best Protocols Section under the Resources tab online.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Phospho-STAT6 (Y641)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P42226, P52633
Signal Transducers and Activators of Transcription 6, IL4 STAT, Interleukin 4 Induced
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
HumanĂ¾Murine
Flow Cytometry
CHI2S4N
PE
STAT6
Conjugated
STAT6
IgG2a, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
6778
Videos

For Research Use Only.