missing translation for 'onlineSavingsMsg'
Learn More

Phospho-STAT5 (Tyr694), FITC, clone: SRBCZX, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-9010-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15566606

  • £191.71 / 100 tests


EU Stock 3
Add to basket

Description

Description

Description: This SRBCZX monoclonal antibody recognizes signal transducer and activator of transcription 5 (STAT5) when phosphorylated on tyrosine 694. STAT proteins are activated by ligand binding to receptors, such as cytokine receptors, that associate with Janus kinase (JAK) family members. Following their phosphorylation by JAKs, STAT proteins translocate to the nucleus where they bind to DNA and regulate transcription of specific genes in a cell type- and cytokine-specific manner. In response to cytokines that signal through the common gamma chain such as IL-2, IL-7, and IL-15, STAT5 is phosphorylated on tyrosine 694 by JAK1 and JAK3. Cytokines such as IL-3, IL-5, and GM-CSF that signal via the common beta chain induce STAT5 phosphorylation on tyrosine 694 by JAK 2. Phosphorylation of STAT5 on tyrosine 694 is essential for STAT5 dimer formation, nuclear translocation, and DNA binding activity. Specificity of this SRBCZX clone was determined by ELISA and flow cytometry.Applications Reported: This SRBCZX antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This SRBCZX antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of stimulated U937 cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Staining Protocol: We recommend using Protocol C: Two-step protocol: Fixation/Methanol. Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins and Protocol B: One-step protocol: intracellular (nuclear) proteins cannot be used. All Protocols can be found in the Flow Cytometry Protocols: Staining Intracellular Antigens for Flow Cytometry Protocol located in the Best Protocols Section under the Resources tab online.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Phospho-STAT5 (Y694)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P42230, P42229
MGF, STAT5
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
HumanĂ¾Murine
Flow Cytometry
SRBCZX
FITC
STAT5A
Conjugated
Stat5a
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
6776
Videos

For Research Use Only.