Learn More
Phospho-S6 (Ser235, Ser236), PerCP-eFluor™ 710, clone: cupk43k, eBioscience™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 46-9007-41
Code : NEW
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15549386
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
For Research Use Only.
Description
Description
This cupk43k monoclonal antibody recognizes human and mouse ribosomal protein S6 (also known as 40S ribosomal protein S6, phosphoprotein NP33, RPS6, RS6, S6) when phosphorylated on serine 235 (S235, human) and serine 236 (S236, mouse). Ribosomal protein S6 is a component of the 40S subunit of the ribosome and is phosphorylated at multiple sites following stimulation of cells by growth factors, tumor promoting agents, or mitogens. Phosphorylation of ribosomal protein S6 by p70S6K and PKDCD results in upregulation of the translation of RNA coding for proteins involved in cell cycle entry. Ribosomal protein S6 is dephosphorylated upon growth arrest.
The specificity of the cupk43k monoclonal antibody was determined by western blotting.
Specifications
Specifications
Phospho-S6 Ribosomal Proteinl (S235/S236)
cupk43k
PerCP-eFluor 710
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
Flow Cytometry (Intracellular Staining)
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
40S ribosomal protein S6, Phosphoprotein NP33
IgG1
RUO
Primary
Murine
Videos