missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Phospho-RSK (S380) Pan DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  R&D Systems™ DYC889BE

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13434825

  • £1962.45 / 1 set

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA kit
Human, mouse, rat
Biotin
15 x 96-well plate
1 Kit
Videos
Documents