Learn More
Phospho-S6 (Ser240, Ser244) Recombinant Rabbit Monoclonal Antibody (11H14L20), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 701845
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with Monkey, Mouse and Rat. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Phospho-S6 (Ser240, Ser244) Recombinant Monoclonal antibody specifically detects Phospho-S6 (Ser240, Ser244) in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Immunoprecipitation, Western Blot.Specifications
Phospho-S6 (Ser240, Ser244) | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P62753 | |
RPS6 | |
Peptide corresponding to Human RPS6 (aa 237-247) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunoprecipitation, Immunoprecipitation, Western Blot | |
11H14L20 | |
Unconjugated | |
RPS6 | |
(Rp)S6; 40S ribosomal protein S6; air[8]; air8; anon-WO02059370.61; CG10944; CG10944-PA; CG10944-PB; Dmel\CG10944; Dmel_CG10944; DS6; hemocytes enlarged; hen; l(1)air[8]; l(1)air8; l(1)hen; LD31286p; lethal(1)aberrant immune response[8]; lethal-(1)-haemocytes enlarged; lethal-aberrant-immune-response-8; M(1)7BC; M(1)7C; minute; minute (1) 7BC; OK/SW-cl.2; OTTHUMP00000021121; OTTHUMP00000021122; Phosphoprotein NP33; pp30; ribosomal protein S6; Rp S6; RP11-513M16.6; RPS6; RpS6-PA; RpS6-PB; RS6; S6; S6 ribosomal protein; S6R; Small ribosomal subunit protein eS6; wu:fa92e06; wu:fb64g06; zgc:92237 | |
Rabbit | |
Protein A | |
RUO | |
6194 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |