Learn More
Phospho-mTOR (Ser2448) Monoclonal Antibody (MRRBY), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 50-9718-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: This MRRBY monoclonal antibody recognizes human and mouse mammalian target of rapamycin (also known as mTOR, FRAP, RAFT) when phosphorylated on S2448. mTOR is a serine/threonine protein kinase that functions as an ATP and amino acid sensor as well as to balance nutrient availability with cell growth, proliferation, motility, survival, protein synthesis, and transcription. Activated mTOR increases production of enzymes necessary for glycolysis and controls the uptake of glucose and other nutrients. Increased glucose uptake and metabolism helps fulfill the energy needs for mTOR-driven cell growth and proliferation. When sufficient nutrients are available, mTOR transmits a positive signal to p70 S6 kinase and participates in the inactivation of the eIF4E inhibitor, 4E-BP1. mTOR is phosphorylated at S2448 via the PI3 kinase/Akt signaling pathway and is autophosphorylated at Ser2481. Due to its critical role in regulation of cell growth, survival, and metabolism, and because it is often abnormally regulated in tumors, mTOR is under investigation as a potential target for anti-cancer therapy. Applications Reported: This MRRBY antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MRRBY antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test.
Phospho-mTOR (Ser2448) Monoclonal antibody specifically detects Phospho-mTOR (Ser2448) in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Phospho-mTOR (Ser2448) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P42345, Q9JLN9 | |
MTOR | |
Affinity chromatography | |
RUO | |
2475, 56717 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MRRBY | |
eFluor 660 | |
MTOR | |
2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS | |
Mouse | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.