missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Histone H3 (Ser28), eFluor 660, clone: HTA28, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-9124-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15500087

  • £74.49 / 25 tests


EU Stock 1
Add to basket

Description

Description

Description: The HTA28 monoclonal antibody recognizes phosphorylated serine 28 of human, mouse, rat, bovine, and hamster histone H3. This 15kDa protein is a component of eukaryotic chromatin that is involved in forming the nucleosome structure. Histone H3 can be phosphorylated at serine 10 and serine 28. Studies have demonstrated that phosphorylation at serine 28 is mediated by MSK1 following activation by the MAP kinase signaling pathway in response to tumor promoters (e.g., UV and EGF) and oncoproteins (e.g., c-Myc, c-Jun, and c-Fos). Histone H3 serine 28 phosphorylation has been linked to chromosome condensation during mitosis, cell transformation, and regulation of RNA polymerase III transcription machinery.Applications Reported: This HTA28 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This HTA28 antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of nocodazole-treated HeLa cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Protocols: We recommend using Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins or Protocol C: Two-step protocol: Fixation/Methanol. Protocol B: One-step protocol: intracellular (nuclear) proteins cannot be used. All Protocols can be found in the Staining intracellular Antigens for Flow Cytometry Protocol located in the Best Protocols Section under the Resources tab online.eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Phospho-Histone H3 (S28)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P68431, P68433
Histone H3, Histone 1, H3a, Histone H3.1, histone H3/a, HIST1H3A
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
HTA28
eFluor 660
HIST1H3A
Conjugated
HIST1H3A
IgG2a, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
8350
Videos

For Research Use Only.