missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Phospho-GSK-3 beta (S9) DuoSet IC, R&D Systems™

Brand:  R&D Systems™ DYC1590-5

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13444335

  • £578.52 / Pack of 5

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
2932
1 Kit
Five 96-well plates
ELISA Development Kit for phosphorylated intracellular proteins
Human, Mouse, Rat
Videos