missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Phospho-GSK-3 beta (S9) DuoSet IC, R&D Systems™

Brand:  R&D Systems™ DYC1590-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13434335

  • £338.10 / Pack of 2


Estimated Shipment date
from Supplier 06-04-2020
Add to basket

Description

Description

Assay Type: ELISA Development Kit for phosphorylated intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Specifications

Specifications

2 96-well plates
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
2932
Human, Mouse, Rat
Videos
Documents