missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Phospho-GSK-3 alpha/beta (S21/S9) DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  R&D Systems™ DYC2630-5

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13454505

  • £773.22 / Pack of 5


Estimated Shipment date
from Supplier 30-03-2020
Add to basket

Description

Description

The Phospho-GSK-3 alpha/beta (S21/S9) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat GSK-3 alpha/beta.
Specifications

Specifications

ELISA
1 Kit
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Human, Mouse, Rat
5 96-well plates
Biotin
ELISA Kit
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Videos
Documents