missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Phospho-ERK1 (T202/Y204)/ERK2 (T185/Y187) DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  R&D Systems™ DYC1018BE

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13464255

  • £1824.95 / Pack of 15


Estimated Shipment
04-04-2019
Add to basket

Description

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA kit
Human, mouse, rat
469.00 - 30000pg/mL
Biotin
15 x 96-well plate
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos