missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Phospho-ERK1 (T202/Y204)/ERK2 (T185/Y187) DuoSet IC ELISA
Phospho-ERK1 (T202/Y204)/ERK2 (T185/Y187) DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 469.00 to 30000 pg/mL |
---|---|
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | ELISA Kit |
Conjugate | Biotin |
Species | Human, Mouse, Rat |
Description
The Phospho-ERK1 (T202/Y204)/ERK2 (T185/Y187) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat ERK1/ERK2.Specifications
469.00 to 30000 pg/mL | |
ELISA Kit | |
Human, Mouse, Rat |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
Biotin |