missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-BACE1 (Ser498) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564480
Additional Details : Weight : 0.01000kg
Description
Phospho-BACE1 (Ser498) Polyclonal Antibody detects endogenous levels of BACE1 only when phosphorylated at Ser498.
Phospho-BACE1 (Ser498) Polyclonal antibody specifically detects Phospho-BACE1 (Ser498) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Phospho-BACE1 (Ser498) | |
Polyclonal | |
Unconjugated | |
BACE1 | |
APP beta-secretase; asp 2; ASP2; Aspartyl protease 2; BACE; BACE 1; BACE1; BACE-1D; BACE-I-432; Beta secretase; Beta-secretase 1; beta-secretase 1 precursor variant 1; beta-site amyloid beta A4 precursor protein-cleaving enzyme; beta-site amyloid precursor protein cleaving enzyme 1; Beta-site APP cleaving enzyme 1; beta-site APP-cleaving enzyme 1; C76936; HSPC104; hypothetical protein LOC403005; KIAA1149; Memapsin; Memapsin2; Memapsin-2; membrane-associated aspartic protease 2; protease; transmembrane aspartic proteinase Asp2; zgc:77409 | |
Rabbit | |
Affinity chromatography | |
RUO | |
23621, 23821, 29392 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P56817, P56818, P56819 | |
BACE1 | |
A synthesized peptide derived from human BACE (Phospho-Ser498) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |