missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-AKT1 (Tyr326) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540167
Code : NEW
Additional Details : Weight : 0.00910kg
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Phospho-AKT1 (Tyr326) Polyclonal antibody specifically detects Phospho-AKT1 (Tyr326) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Phospho-AKT1 (Tyr326) | |
Polyclonal | |
Unconjugated | |
AKT1 | |
Akt; Akt kinase; AKT serine/threonine kinase 1; Akt/PKB; akt1; AKT1 kinase; AKT1m; Akt1m protein; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; CAKT; C-AKT; CG4006; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; CWS6; dAkt; D-Akt; dAkt kinase; dAKT/dPKB; dAkt/PKB; DAKT1; DAKT1/PKB; Dmel\CG4006; Dmel_CG4006; dPKB; DRAC-PK; DRAC-PK66; DRAC-PK85; l(3)04226; l(3)89Bq; murine thymoma viral (v-akt) oncogene homolog 1; pAkt; P-AKT; PKB; PKB alpha; PKB beta; PKB gamma; PKB/AKT; PKB/dAKT; PKBalpha; PKB-ALPHA; PKBG; PRKBA; protein kinase B; Protein kinase B alpha; protein kinase B-alpha; proto-oncogene c-Akt; RAC; rac protein kinase alpha; RAC protein kinase alpha RAC-PK alpha; RAC serine/threonine protein kinase; RAC serine/threonine-protein kinase; RAC-ALPHA; RAC-alpha serine/threonine-protein kinase; RacPK; RAC-PK-alpha; RAC-PK-beta; RAC-PK-gamma; related to A and C kinases; related to PKA to PKC protein kinases; related to the A and C kinases; SD10374p; serine/threonine protein kinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11651, 207, 24185 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P31749, P31750, P47196 | |
AKT1 | |
Peptide sequence around phosphorylation site of tyrosine 326 (N-D-Y(p)-G-R) derived from Human Akt. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |