missing translation for 'onlineSavingsMsg'
Learn More

Phospho-AKT1 (Ser473) Mouse anti-Human, Mouse, FITC, Clone: SDRNR, eBioscience™

Mouse Monoclonal Antibody

£78.75 - £206.00

Specifications

Antigen Phospho-AKT1 (Ser473)
Clone SDRNR
Target Species Human, Mouse
Conjugate FITC
Isotype IgG2a, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15576626
View Documents
Affymetrix eBioscience
11-9715-41
25 Tests £78.75
25 tests View alternative products
Save Up to 
»
Estimated Shipment
30-11-2020
Please to purchase this item. Need a web account? Register with us today!
15586626
View Documents
Affymetrix eBioscience
11-9715-42
100 Tests £206.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment
05-11-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Staining Protocol: All protocols work well for this monoclonal antibody. Use of Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins allows for the greatest flexibility for detection of surface and intracellular (cytoplasmic) proteins. Use of Protocol B: One-step protocol: intracellular (nuclear) proteins is recommended for staining of transcription factors in conjunction with surface and phosphorylated intracellular (cytoplasmic) proteins. Protocol C: Two-step protocol: Fixation/Methanol allows for the greatest discrimination of phospho-specific signaling between unstimulated and stimulated samples, but with limitations on the ability to stain specific surface proteins (refer to "Clone Performance Following Fixation/Permeabilization" located in the Best Protocols Section under the Resources tab online). All Protocols can be found in the Flow Cytometry Protocols: "Staining Intracellular Antigens for Flow Cytometry Protocol" located in the Best Protocols Section under the Resources tab online. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. AKT also known as protein kinase B (PKB) or RAS-alpha, is an ubiquitous serine/threonine kinase that plays an important role in diverse biological responses such as regulation of metabolism, cell survival and growth by phosphorylating multiple proteins. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.
Specifications

Specifications

Phospho-AKT1 (Ser473)
Human, Mouse
IgG2a, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
SDRNR
FITC
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.