missing translation for 'onlineSavingsMsg'
Learn More

Phospho-AKT1 (Ser473) Mouse anti-Human, Mouse, FITC, Clone: SDRNR, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-9715-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15586626

  • £206.00 / 100 tests

Estimated Shipment
30-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Staining Protocol: All protocols work well for this monoclonal antibody. Use of Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins allows for the greatest flexibility for detection of surface and intracellular (cytoplasmic) proteins. Use of Protocol B: One-step protocol: intracellular (nuclear) proteins is recommended for staining of transcription factors in conjunction with surface and phosphorylated intracellular (cytoplasmic) proteins. Protocol C: Two-step protocol: Fixation/Methanol allows for the greatest discrimination of phospho-specific signaling between unstimulated and stimulated samples, but with limitations on the ability to stain specific surface proteins (refer to "Clone Performance Following Fixation/Permeabilization" located in the Best Protocols Section under the Resources tab online). All Protocols can be found in the Flow Cytometry Protocols: "Staining Intracellular Antigens for Flow Cytometry Protocol" located in the Best Protocols Section under the Resources tab online. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. AKT also known as protein kinase B (PKB) or RAS-alpha, is an ubiquitous serine/threonine kinase that plays an important role in diverse biological responses such as regulation of metabolism, cell survival and growth by phosphorylating multiple proteins. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.
Specifications

Specifications

Phospho-AKT1 (Ser473)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P31750, P31749
AKT, MGC99656, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry
SDRNR
FITC
AKT1
Liquid
Akt1
IgG2a, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
11651, 207
Videos
Documents

For Research Use Only.