missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKT (Phospho) [pS473] Multispecies InstantOne™ ELISA Kit
InstantOne ELISA
£460.00 - £19070.00
Specifications
For Use With (Application) | ELISA |
---|---|
Storage Requirements | 2°C to 8°C |
Synonym | PKB, Protein Kinase B, RAC-PK-alpha |
Alert:
Please contact Technical Service for a lot specific COA
Includes: User Manual
InstantOne™ Assay Plate - 96 well (sealed)
Cell Lysis Buffer (5X)
Enhancer Solution
Wash Buffer (10X)
phospho-AKT 1/2/3 (Ser473) Capture Antibody Reagent
phospho-AKT 1/2/3 (Ser473) Detection Antibody Reagent
Positive Control Cell Lysate (lyophilized)
Detection Reagent (TMB Substrate)
Stop Solution
Plate seal
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15540777
|
Invitrogen™
85-86042-11 |
96 Tests |
£460.00
96 tests |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15530777
|
Invitrogen™
85-86042-103 |
10 x 384 Tests |
£5750.00
Pack of 10 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15550777
|
Invitrogen™
85-86042-253 |
25 x 384 Tests |
£11490.00
Pack of 25 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15560777
|
Invitrogen™
85-86042-503 |
50 x 384 Tests |
£19070.00
Pack of 50 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human, mouse, and hamster AKT 1/2/3 in cell lysates. Detection of rat AKT 1/2/3 is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.Specifications
ELISA | |
2°C to 8°C | |
PKB, Protein Kinase B, RAC-PK-alpha |