Alfa Aesar™ Phosphate-buffered RIPA


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J62694.AP

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.00050kg



Product Code. 15465019

Quantity Price
1 £ 55.65 / 500mL
Estimated Shipment date
from Supplier 10-05-2017
Add to basket


Description and Specification

Specification

Quantity 500mL
Storage Requirements 2° to 8°C
Product Type Cell Lysis Reagent
Format Liquid
Documentation

Technical documents