missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Phosphate-buffered RIPA, 2X

Brand:  Alfa Aesar™ J60780.AK

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15443489

  • £55.65 / 250mL


Estimated Shipment date
from Supplier 10-08-2017
Add to basket

Specifications Description & Specifications

Specifications

Colorless
Liquid
2° to 8°C
250mL
Cell Lysis Reagent
2X
Documentation

Technical Documents