missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer Solution, pH 7, Honeywell™ Fluka™

Catalog Number p-7120665
Click to view available options
Quantity:
1 L
10 L
Packaging:
Poly bottle
Poly pail
This item is not returnable. View return policy
7732-18-5
This item is not returnable. View return policy
7732-18-5
Density 1.01g/mL at 20°C
Quantity 1 L
Packaging Poly bottle
UN Number NONH for all modes of transport
Chemical Name or Material Phosphate buffer solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'