missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Phosphatase Inhibitor Cocktail B

For the inhibition of protein tyrosine phosphatases, acid- and alkaline-phosphatases

Brand:  Alfa Aesar™ J64651.LQ

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 2811 Chem class : 1 Pack group : III

 View more versions of this product

Product Code. 15446499

  • £41.23 / 1 unit


Estimated Shipment date
from Supplier 13-02-2018
Add to basket

Specifications

Specifications

For the inhibition of protein tyrosine phosphatases, acid- and alkaline-phosphatases
Lyophilized Solid
−30° to −10°C
Hygroscopic
White
Phosphatase Inhibitor Cocktail
1 Unit
Videos
SDS