Learn More
Phosphatase buffer 1 (5X), Thermo Scientific™
For cellular and molecular biology applications
Brand: Thermo Scientific Alfa Aesar J62267.AK
Code : NEW
Additional Details : Weight : 0.00025kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Formulation: 100mM Tris (pH 7.5) and 100mM magnesium chloride
Specifications
100mM Tris (pH 7.5) and 100mM magnesium chloride | |
250 mL | |
Liquid |
Ambient temperatures | |
Phosphatase buffer 1 (5X) |
For Research Use Only