missing translation for 'onlineSavingsMsg'
Learn More

Phorbol 12-myristate 13-acetate, Tocris Bioscience™

Protein kinase C activator

Brand:  Tocris (Bio-Techne) 12015

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 5mg

 View more versions of this product

Product Code. 13458853

  • £229.69 / 5mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Extensively used phorbol ester activator of protein kinase C; tumor promoter. Binds to PKC with a Ki value of 2.6 nM as measured by displacement of [3H]phorbol 12,13-dibutyrate binding in rat cortex synaptosomal membranes.
Specifications

Chemical Identifiers

16561-29-8
616.836
phorbol 12-myristate 13-acetate, phorbol ester, 12-o-tetradecanoylphorbol-13-acetate, 12-o-tetradecanoylphorbol 13-acetate, factor a1, tetradecanoylphorbol acetate, 12-tetradecanoylphorbol 13-acetate, factor a1 croton oil, phorbol 13-acetate 12-myristate, phorbol-12-myristate-13-acetate
CHEBI:37537
C36H56O8
PHEDXBVPIONUQT-RGYGYFBISA-N
27924
CCCCCCCCCCCCCC(=O)OC1C(C2(C(C=C(CC3(C2C=C(C3=O)C)O)CO)C4C1(C4(C)C)OC(=O)C)O)C

Specifications

Phorbol 12-myristate 13-acetate
16561-29-8
phorbol 12-myristate 13-acetate, phorbol ester, 12-o-tetradecanoylphorbol-13-acetate, 12-o-tetradecanoylphorbol 13-acetate, factor a1, tetradecanoylphorbol acetate, 12-tetradecanoylphorbol 13-acetate, factor a1 croton oil, phorbol 13-acetate 12-myristate, phorbol-12-myristate-13-acetate
CCCCCCCCCCCCCC(=O)OC1C(C2(C(C=C(CC3(C2C=C(C3=O)C)O)CO)C4C1(C4(C)C)OC(=O)C)O)C
27924
616.83
5mg
C36H56O8
PHEDXBVPIONUQT-RGYGYFBISA-N
616.836
CHEBI:37537
>99%
Videos