Learn More
Grant Instruments™ PMHT Thermoshaker for Microtubes and Microplates
Accelerates in 3 seconds for fast start but also operates smoothly and quietly, this Grant Instruments™ PMHT Thermoshaker for Microtubes and Microplates is ideal for DNA extraction, enzyme reactions and cellular biology techniques.
Brand:  Grant Instruments™ PHMT-PSC24N
Code : NEW
Additional Details : Weight : 4.00000kg
 View more versions of this product
Product Code. 15281166
  • £1206.00 / Each
Estimated Shipment
27-02-2018
Add to basket
Description
Description
  • Combines three instruments in one:
    • Microtube and microplate thermoshaker
    • Compact benchtop incubator
    • Microtube and microplate shaker in cold or ambient temperatures
  • 2-line LCD display clearly indicates both set and actual values for temperature, shaking speed and time.
  • Interchangeable block for 20 or 12 microtubes provides flexibility for an easy change in application.
  • Operates extremely smoothly, quietly and consistently.
  • Includes ‘soft’ start function for delicate samples.
  • Fast start - acceleration 3 seconds.
  • Compact and sturdy, with a low profile and small footprint.
Specifications
Specifications
PHMT-PSC24N
+5°C above ambient to 100°C
0.1°C at 37°C; 0.2°C at 60°C; 0.2°C at 100°C
24 x 1.5mL microtubes
2 line x 16 character LCD
205mm
250 to 1400rpm (increments 10rpm)
+25 to 100°C
2mm
230mm
130mm
4kg
Videos