missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Philips Glass Beakers With Spout

Brand:  Duran™ 211412904

Code : A0

Additional Details : Weight : 0.70000kg

 View more versions of this product

Product Code. 11776808

  • £245.00 / Pack of 10


Estimated Shipment
27-06-2018
Add to basket

Specifications

Specifications

Clear
87mm
Beaker
Borosilicate Glass
59mm
10/Pk.
150mL
Videos