missing translation for 'onlineSavingsMsg'
Learn More

Philanthotoxin 74, Tocris Bioscience™

GluA1 and GluA3 selective AMPA antagonist

Brand:  Tocris (Bio-Techne) 2770/1

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 1mg

 View more versions of this product

Product Code. 13430564

  • £91.14 / 1mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

AMPA receptor antagonist. Non-selective homomeric GluA3 and GluA1 receptor antagonist (IC50 values are 263 and 296 nM respectively). Inhibits homomeric GluA3 and GluA1 with a 100-fold greater potency than heteromeric GluA1/2 and GluA2/3 receptors.
Specifications

Chemical Identifiers

1227301-51-0
507.541
philanthotoxin 74, philanthotoxin 74 dihydrochloride, phtx-74, philanthotoxin-7,4, phtx-74 dihydrochloride, n-7-4-aminobutyl amino heptyl-nalpha-butanoyl-l-tyrosinamide-hydrogen chloride 1/2, s-n-7-4-aminobutyl amino heptyl-4-hydroxy-?-1-oxobutyl amino benzenepropanamide dihydrochloride, s-n-7-4-aminobutyl amino heptyl-4-hydroxy-alpha-1-oxobutyl amino benzenepropanamide dihydrochloride, n-1s-1-7-4-aminobutyl amino heptyl carbamoyl-2-4-hydroxyphenyl ethyl butanamide dihydrochloride
N-[(2S)-1-[7-(4-aminobutylamino)heptylamino]-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]butanamide;dihydrochloride
C24H44Cl2N4O3
HWTJQQMIKVJWLH-IKXQUJFKSA-N
46213501
CCCC(=O)NC(CC1=CC=C(C=C1)O)C(=O)NCCCCCCCNCCCCN.Cl.Cl

Specifications

1mg
1227301-51-0
philanthotoxin 74, philanthotoxin 74 dihydrochloride, phtx-74, philanthotoxin-7,4, phtx-74 dihydrochloride, n-7-4-aminobutyl amino heptyl-nalpha-butanoyl-l-tyrosinamide-hydrogen chloride 1/2, s-n-7-4-aminobutyl amino heptyl-4-hydroxy-?-1-oxobutyl amino benzenepropanamide dihydrochloride, s-n-7-4-aminobutyl amino heptyl-4-hydroxy-alpha-1-oxobutyl amino benzenepropanamide dihydrochloride, n-1s-1-7-4-aminobutyl amino heptyl carbamoyl-2-4-hydroxyphenyl ethyl butanamide dihydrochloride
CCCC(=O)NC(CC1=CC=C(C=C1)O)C(=O)NCCCCCCCNCCCCN.Cl.Cl
507.541
507.54
Philanthotoxin 74
C24H44Cl2N4O3
HWTJQQMIKVJWLH-IKXQUJFKSA-N
N-[(2S)-1-[7-(4-aminobutylamino)heptylamino]-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]butanamide;dihydrochloride
46213501
>98%
Videos
Documents