missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylammonium Iodide 99%, ACROS Organics™

Brand:  Acros Organics 160685000

Code : 29

Additional Details : CAS Number : 98-04-4 Weight : 0.55000kg


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10265730

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Phase transfer catalyst
Specifications

Chemical Identifiers

Phenyltrimethylammonium iodide
98-04-4
99%
C6H5N(CH3)3I
MFCD00011791
99%
≥98.5%
C9H14IN
263.11
(N, N, N)-Trimethylanilinium iodide, Trimethylphenylammonium iodide

Specifications

Phenyltrimethylammonium iodide
500g
98-04-4
Glass bottle
≥98.5%
C9H14IN
263.11
15, 7428
White to slightly green
99%
Authentic
Slightly Crystalline Powder or Flakes
99%
C6H5N(CH3)3I
MFCD00011791
(N, N, N)-Trimethylanilinium iodide, Trimethylphenylammonium iodide
Videos
SDS