missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethoxysilane, 85%, ACROS Organics™

Brand:  Acros Organics 370642500

Code : 29

Additional Details : CAS Number : 2996-92-1 Weight : 0.26550kg Transport : UN number : 1993 Chem class : 3 Pack group : III


Packaging Glass bottle
Quantity 250mL

 View more versions of this product

Product Code. 10603583

  • £24.70 / 250mL


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

Phenyltrimethoxysilane
85%
C6H5Si(OCH3)3
MFCD00025689
≥80%
C9H14O3Si
198.29
Trimethoxysilylbenzene

Specifications

Phenyltrimethoxysilane
Clear colorless
2996-92-1
Glass bottle
Liquid
85%
C6H5Si(OCH3)3
MFCD00025689
Trimethoxysilylbenzene
1.0620g/mL
250mL
Authentic
1.4730 to 1.4750 (20°C, 589nm)
≥80%
C9H14O3Si
198.29
16, IV, 1556
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Phenyltrimethoxysilane

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS