missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyltin trichloride, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Phenyltin trichloride is used to produce 3-phenyl-cyclohexanone with cyclohex-2-enone. And it is also used in pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.84 g/mL |
Odor | Pungent |
Quantity | 5 g |
UN Number | UN2788 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 302.18 |
Physical Form | Liquid |
Chemical Name or Material | Phenyltin trichloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'