missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenyl bromide, 98%, ACROS Organics™

Brand:  Acros Organics 223060250

Code : 29

Additional Details : CAS Number : 34837-55-3 Weight : 0.07500kg Transport : UN number : 2928 Chem class : 6.1 Pack group : II

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10713421

  • £123.40 / 25g


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

Phenylselenyl bromide
34837-55-3
98%
C6H5SeBr
MFCD00000047
98%
≥97.5%
C6H5BrSe
235.97
Benzeneselenyl bromide

Specifications

Phenylselenyl bromide
25g
34837-55-3
Glass bottle
≥97.5%
C6H5BrSe
235.97
06, II, 319
Benzeneselenyl bromide
Dark brownish to black
98%
Authentic
Crystals
98%
C6H5SeBr
MFCD00000047
06,459; 07,286; 11,33; 13,26; 01,1047; 11,487; 16,272
Videos
SDS