missing translation for 'onlineSavingsMsg'
Learn More

Phenylpropiolic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-183681
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
637-44-5
C9H6O2
146.15
MFCD00004361
XNERWVPQCYSMLC-UHFFFAOYSA-N
phenylpropiolic acid, 3-phenylpropiolic acid, phenylpropynoic acid, 3-phenylpropynoic acid, 2-propynoic acid, 3-phenyl, phenylacetylenecarboxylic acid, 3-phenyl-2-propynoic acid, phenylacetylene monocarboxylic acid, propiolic acid, 3-phenyl, phenyl propiolic acid
69475
3-phenylprop-2-ynoic acid
OC(=O)C#CC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
637-44-5
C9H6O2
146.15
MFCD00004361
XNERWVPQCYSMLC-UHFFFAOYSA-N
phenylpropiolic acid, 3-phenylpropiolic acid, phenylpropynoic acid, 3-phenylpropynoic acid, 2-propynoic acid, 3-phenyl, phenylacetylenecarboxylic acid, 3-phenyl-2-propynoic acid, phenylacetylene monocarboxylic acid, propiolic acid, 3-phenyl, phenyl propiolic acid
69475
3-phenylprop-2-ynoic acid
OC(=O)C#CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 136.0°C to 139.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5C≡CCO2H
Quantity 5 g
Beilstein 09, 633
Solubility Information Solubility in water: freely soluble
Formula Weight 146.15
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phenylpropiolic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Phenylpropiolic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'