missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylphosphonic dichloride, 90+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
Phenylphosphonic dichloride is used in the preparation of 2-phenyl-[1,3,2]dioxaphosphepane-2-oxide. Further, it acts as an effective reagent for the chlorinative dehydration of some heterocycles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenylphosphonic dichloride is used in the preparation of 2-phenyl-[1,3,2]dioxaphosphepane-2-oxide. Further, it acts as an effective reagent for the chlorinative dehydration of some heterocycles.
Solubility
Miscible with benzene, chloroform, dimethyl sulfoxide and carbon tetrachloride.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong bases and water.
Phenylphosphonic dichloride is used in the preparation of 2-phenyl-[1,3,2]dioxaphosphepane-2-oxide. Further, it acts as an effective reagent for the chlorinative dehydration of some heterocycles.
Solubility
Miscible with benzene, chloroform, dimethyl sulfoxide and carbon tetrachloride.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong bases and water.
Specifications
Melting Point | 1°C to 3°C |
Density | 1.379 |
Boiling Point | 258°C |
Flash Point | 204°C (399°F) |
Refractive Index | 1.56 |
Quantity | 50 g |
UN Number | UN3265 |
Beilstein | 1072240 |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible with benzene,chloroform,dimethyl sulfoxide and carbon tetrachloride. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'