missing translation for 'onlineSavingsMsg'
Learn More

Phenylmercury(II) chloride, 96%, Hg 63.5%, Thermo Scientific Chemicals

Catalog Number p-7050420
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
100-56-1
C6H5ClHg
313.15
MFCD00000463
AWGTVRDHKJQFAX-UHFFFAOYSA-M
phenylmercuric chloride, agrenal, phenylmercury chloride, mercury, chlorophenyl, hexason, mercutal, merfazin, stopspot, agronal, mersolite 2
7511
chloro(phenyl)mercury
Cl[Hg]C1=CC=CC=C1
This item is not returnable. View return policy
100-56-1
C6H5ClHg
313.15
MFCD00000463
AWGTVRDHKJQFAX-UHFFFAOYSA-M
phenylmercuric chloride, agrenal, phenylmercury chloride, mercury, chlorophenyl, hexason, mercutal, merfazin, stopspot, agronal, mersolite 2
7511
chloro(phenyl)mercury
Cl[Hg]C1=CC=CC=C1

The reaction of phenylmercury (II) chloride with acetophenonethiosemicarbazone (Hatsc) in ethanol in 1: 1 mole ratio undergoes symmetrisation forming the products, HgCl2 (Hatse) 2 and Ph2Hg instead of the anticipated compound PhHgCl (Hatse). A new method for the determination of chloride ion is based on the formation of phenylmercury(II) chloride, its extraction into chloroform and reaction with sodium diethyldithiocarbamate to form phenylmercury(II) diethyldithiocarbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 246°C to 250°C
Color White
Quantity 10 g
UN Number UN2026
Merck Index 14,7301
Solubility Information Soluble in benzene, ether, pyridine; Very slightly soluble in water
Formula Weight 313.18
Percent Purity 96%
Concentration or Composition (by Analyte or Components) Hg 63.5%
Physical Form Powder
Chemical Name or Material Phenylmercury(II) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'