Learn More
Phenylmercury(II) chloride, 96%, Hg 63.5%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
The reaction of phenylmercury (II) chloride with acetophenonethiosemicarbazone (Hatsc) in ethanol in 1: 1 mole ratio undergoes symmetrisation forming the products, HgCl2 (Hatse) 2 and Ph2Hg instead of the anticipated compound PhHgCl (Hatse). A new method for the determination of chloride ion is based on the formation of phenylmercury(II) chloride, its extraction into chloroform and reaction with sodium diethyldithiocarbamate to form phenylmercury(II) diethyldithiocarbamate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 246°C to 250°C |
Color | White |
Quantity | 50 g |
UN Number | UN2026 |
Merck Index | 14,7301 |
Solubility Information | Soluble in benzene, ether, pyridine; Very slightly soluble in water |
Formula Weight | 313.18 |
Percent Purity | 96% |
Concentration or Composition (by Analyte or Components) | Hg 63.5% |
Physical Form | Powder |
Show More |
RUO – Research Use Only