Learn More
Phenylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Grignard reagent Phenylmagnesium bromide is an important reagent called as Grignard reagent used in the synthesis of organic and oraganometallic reactions. It is used to prepare phenyl aryl sulphides, sulphoxides, and sulphones. It acts as a strong acid and Lewis acid and undergoes addition reaction with many unsaturated functional groups. It is involved in the synthesis of end-functionalized regioregular poly(3-alkylthiophene)s, 1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol, o-substituted benzophenones and 6-benzyl-1,3,3-trimethyl-2-oxabicyclo[2.2.2]octan-6-ol. In addition to this, it is used for the monalkylation of primary amines to get secondary amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow to Brown |
Density | 1.14 g/mL |
Odor | Ether-like |
Quantity | 100 mL |
UN Number | UN3399 |
Sensitivity | Air, light, and moisture sensitive; May form a Precipitate |
Solubility Information | Miscible with terahydrofuran. |
IUPAC Name | bromo(phenyl)magnesium |
Formula Weight | 181.31 |
Concentration or Composition (by Analyte or Components) | 3M in ether |
Show More |
RUO – Research Use Only