missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Phenyllithium, typically 1.9M in di-n-butyl ether

Brand:  Alfa Aesar™ 045770.6C

Code : NEW

Additional Details : CAS Number : 591-51-5 Weight : 0.00010kg Transport : UN number : 2924 Chem class : 1 Pack group : II

Quantity 5 x 100mL

 View more versions of this product

Product Code. 15427065

  • £230.02 / 100mL


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

Phenyllithium
C6H5Li
MFCD00014254
506502
591-51-5
84.05
UN2924

Specifications

Phenyllithium
591-51-5
0.81g/mL
Dark brown to black
Air & Moisture Sensitive
C6H5Li
MFCD00014254
506502
Typically 1.9M in di-n-butyl ether
5 x 100mL
140° to 143°C
25°C (77°F)
Liquid
84.05
UN2924
Videos
SDS