missing translation for 'onlineSavingsMsg'
Learn More

Phenylglyoxal monohydrate, 97%, Thermo Scientific Chemicals

Catalog Number 10700861
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1075-06-5
C8H6O2·H2O
152.15
MFCD00149499
NBIBDIKAOBCFJN-UHFFFAOYSA-N
dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone
99611
2,2-dihydroxy-1-phenylethanone
C1=CC=C(C=C1)C(=O)C(O)O
This item is not returnable. View return policy
1075-06-5
C8H6O2·H2O
152.15
MFCD00149499
NBIBDIKAOBCFJN-UHFFFAOYSA-N
dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone
99611
2,2-dihydroxy-1-phenylethanone
C1=CC=C(C=C1)C(=O)C(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 71.0°C to 76.0°C
Color Pink or White to Yellow
Boiling Point 142.0°C (125.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5COCHO·H2O
Quantity 25 g
Beilstein 07, 670
Formula Weight 152.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Phenylglyoxal monohydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'