missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylglyoxal monohydrate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.
Solubility
Partly miscible with water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.
Solubility
Partly miscible with water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 71°C to 77°C |
Boiling Point | 142°C (125 mmHg) |
Quantity | 5 g |
Beilstein | 1854721 |
Solubility Information | Partly miscible with water. |
Formula Weight | 152.15 (134.12 Anhydrous) |
Percent Purity | 97% |
Chemical Name or Material | Phenylglyoxal monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'