missing translation for 'onlineSavingsMsg'
Learn More

Phenylethyl isothiocyanate, 96%, Thermo Scientific Chemicals

Catalog Number p-182832
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2257-09-2
C9H9NS
163.24
MFCD00004821
IZJDOKYDEWTZSO-UHFFFAOYSA-N
This item is not returnable. View return policy
2257-09-2
C9H9NS
163.24
MFCD00004821
IZJDOKYDEWTZSO-UHFFFAOYSA-N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 113.0°C (1.0 mmHg)
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula C6H5CH2CH2NCS
Refractive Index 1.5860 to 1.5910
Quantity 5 g
Beilstein 12, 1100
Merck Index 15, 7340
Specific Gravity 1.1
Synonym 2,6-dichlorofluorobenzene, 1,3-dichloro-2-fluoro-benzene, 2,6-dichloro-1-fluorobenzene, 2,6-dichloro fluoro benzene, benzene,1,3-dichloro-2-fluoro, benzene, 1,3-dichloro-2-fluoro, pubchem2311, 2,6-dichlorfluorbenzol, acmc-2097ae, 2,6-dichloro fluorobenzene
Solubility Information Solubility in water: insoluble
SMILES S=C=NCCC1=CC=CC=C1
IUPAC Name 1,3-dichloro-2-fluorobenzene
PubChem CID 137520
Formula Weight 163.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Phenylethyl isothiocyanate, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenylethyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'