missing translation for 'onlineSavingsMsg'
Learn More

Phenylazoformic acid 2-phenylhydrazide, suitable for colorimetric analysis, Thermo Scientific Chemicals

Catalog Number 10776391
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
538-62-5
C13H12N4O
240.26
MFCD00003024
ZFWAHZCOKGWUIT-UHFFFAOYSA-N
1,5-diphenylcarbazone, diazenecarboxylic acid, phenyl-, 2-phenylhydrazide, unii-5uyj4r0d5t, phenylazoformic acid 2-phenylhydrazide, sym-diphenylcarbazone, 5uyj4r0d5t, chembl79409, 3-hydroxy-1,5-diphenylformazan, phenyldiazenecarboxylic acid 2-phenylhydrazide, 3-phenylamino-1-phenylimino urea
10860
1-anilino-3-phenyliminourea
C1=CC=C(C=C1)NNC(=O)N=NC2=CC=CC=C2
This item is not returnable. View return policy
538-62-5
C13H12N4O
240.26
MFCD00003024
ZFWAHZCOKGWUIT-UHFFFAOYSA-N
1,5-diphenylcarbazone, diazenecarboxylic acid, phenyl-, 2-phenylhydrazide, unii-5uyj4r0d5t, phenylazoformic acid 2-phenylhydrazide, sym-diphenylcarbazone, 5uyj4r0d5t, chembl79409, 3-hydroxy-1,5-diphenylformazan, phenyldiazenecarboxylic acid 2-phenylhydrazide, 3-phenylamino-1-phenylimino urea
10860
1-anilino-3-phenyliminourea
C1=CC=C(C=C1)NNC(=O)N=NC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Melting Point 153.0°C to 157.0°C
Color Orange
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5N=NCONHNHC6H5
Quantity 10 g
Beilstein 16,24
Merck Index 15,3353
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,chloroform and benzene
Formula Weight 240.26
Physical Form Powder
Chemical Name or Material Phenylazoformic acid 2-phenylhydrazide, suitable for colorimetric analysis
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'