missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsonic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7080893
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
98-05-5
C6H7AsO3
202.041
MFCD00002097
LVKZSFMYNWRPJX-UHFFFAOYSA-N
benzenearsonic acid, monophenylarsonic acid, arsonic acid, phenyl, unii-57f9ku116m, kyselina benzenarsonova czech, arsonic acid, as-phenyl, kyselina benzenarsonova, benzenarsonic acid, benzenearsonie acid, acmc-20ajwq
7365
CHEBI:29851
phenylarsonic acid
C1=CC=C(C=C1)[As](=O)(O)O
This item is not returnable. View return policy
98-05-5
C6H7AsO3
202.041
MFCD00002097
LVKZSFMYNWRPJX-UHFFFAOYSA-N
benzenearsonic acid, monophenylarsonic acid, arsonic acid, phenyl, unii-57f9ku116m, kyselina benzenarsonova czech, arsonic acid, as-phenyl, kyselina benzenarsonova, benzenarsonic acid, benzenearsonie acid, acmc-20ajwq
7365
CHEBI:29851
phenylarsonic acid
C1=CC=C(C=C1)[As](=O)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 158°C to 162°C (decomposition)
Quantity 5 g
UN Number UN3465
Merck Index 14,1067
Formula Weight 202.04
Percent Purity 97%
Chemical Name or Material Phenylarsonic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'