missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetylene, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081925
Click to view available options
Quantity:
25 g
100 g
500 g
536-74-3
C8H6
102.136
MFCD00008570
UEXCJVNBTNXOEH-UHFFFAOYSA-N
phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
10821
ethynylbenzene
C#CC1=CC=CC=C1
536-74-3
C8H6
102.136
MFCD00008570
UEXCJVNBTNXOEH-UHFFFAOYSA-N
phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
10821
ethynylbenzene
C#CC1=CC=CC=C1

Phenylacetylene is involved in the preparation of styrene by reduction using Lindlar catalyst. It is used to study the mechanism of the palladium-catalyzed phenyl acetylene oxidative carbonylation reaction. It is also used in the polymerization process to prepare polyphenylacetylene namely 1,2,4-triphenylbenzene and 1,3,5-triphenylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylacetylene is involved in the preparation of styrene by reduction using Lindlar catalyst. It is used to study the mechanism of the palladium-catalyzed phenyl acetylene oxidative carbonylation reaction. It is also used in the polymerization process to prepare polyphenylacetylene namely 1,2,4-triphenylbenzene and 1,3,5-triphenylbenzene.

Solubility
Immiscible with water.

Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, halogens, alkali metals and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -45°C
Density 0.93
Boiling Point 142°C to 144°C
Flash Point 27°C (81°F)
Refractive Index 1.548
Quantity 25 g
UN Number UN3295
Beilstein 605461
Sensitivity Light sensitive
Merck Index 14,3861
Solubility Information Immiscible with water.
Formula Weight 102.14
Percent Purity ≥98%
Chemical Name or Material Phenylacetylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenylacetylene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : ethynyl-benzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'