missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetic acid hydrazide, 98%, Thermo Scientific Chemicals

Catalog Number p-3779403
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
937-39-3
C8H10N2O
150.18
MFCD00007612
FPTCVTJCJMVIDV-UHFFFAOYSA-N
phenylacetic acid hydrazide, benzeneacetic acid, hydrazide, phenylacetic hydrazide, phenylacetylhydrazine, phenylacetyl hydrazide, phenacetic acid hydrazide, 2-phenylacetyl hydrazine, phenylacetohydrazide, phenylacethydrazide, phenyl-acetic acid hydrazide
70301
2-phenylacetohydrazide
C1=CC=C(C=C1)CC(=O)NN
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
937-39-3
C8H10N2O
150.18
MFCD00007612
FPTCVTJCJMVIDV-UHFFFAOYSA-N
phenylacetic acid hydrazide, benzeneacetic acid, hydrazide, phenylacetic hydrazide, phenylacetylhydrazine, phenylacetyl hydrazide, phenacetic acid hydrazide, 2-phenylacetyl hydrazine, phenylacetohydrazide, phenylacethydrazide, phenyl-acetic acid hydrazide
70301
2-phenylacetohydrazide
C1=CC=C(C=C1)CC(=O)NN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 115.0°C to 117.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2CONHNH2
Quantity 25 g
Beilstein 09, 446
Formula Weight 150.18
Percent Purity 98%
Physical Form Needles
Chemical Name or Material Phenylacetic acid hydrazide
Show More Show Less
compliance-icons
Product Identifier
  • Phenylacetic acid hydrazide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'